Diammonium hydrogen orthophosphate ≥99.0% (by acidimetry), EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
Synonyms:
Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate
1.01207.0500 replaces and is the same as AX1355-1. Part # Change only, Same Pack Size, Same Specifications.
1.01207.0500 replaces and is the same as AX1355-3. Different Pack Size, Same Specifications.
1.01207.9050 replaces and is the same as AX1355-20. Different Pack Size, Same Specifications.
Formula:
(NH₄)₂HPO₄ Melting Pt: 185 °C Density: 1.619 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010891 CAS Number: 7783-28-0 EINECS: 231-987-8 REACH: 01-2119490974-22 Merck Index: 13,00546 |
Specification Test Results
Solubility | 690 g/l (20 °C) |
Molar Mass | 132.05 g/mol |
Bulk Density | 800 - 1000 kg/m3 |
Density | 1.619 g/cm3 (20 °C) |
pH | 7.6 - 8.2 (100 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...