Ammonium persulfate (APS), EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.01201.0500
1.01201.1000
1.01201.5000
1.01201.9012
CA1.01201.0500CS
1016
CAD
CA1.01201.0500
CA1.01201.1000
CA1.01201.5000
EM1.01201.9012
Ammonium persulfate (APS), EMSURE® for analysis, Supelco®
Ammonium peroxodisulphate (APS)
1.01201.0500 replaces and is the same as AX1340-1. Part # Change only, Same Pack Size, Same Specifications.
1.01201.5000 replaces and is the same as AX1340-3. Different Pack Size, Same Specifications.
1.01201.9050 replaces and is the same as AX1340-20. Part # Change only, Same Pack Size, Same Specifications.
Formula:
(NH₄)₂S₂O₈ Melting Pt: 120 °C Density: 1.98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003390 CAS Number: 7727-54-0 EINECS: 231-786-5 UN: 1444 ADR: 5.1,III Merck Index: 13,00545 |
|
Specification Test Results
Solubility | 620 g/l (20 °C) |
Melting Point | 120 °C (decomposition) |
Molar Mass | 228.19 g/mol |
Bulk Density | 900 kg/m3 |
Density | 1.98 g/cm3 (20 °C) |
pH | 3.2 (100 g/l, H2O, 20 °C) |
Hazard Class | 5.1 |
Hazard Class Description | Oxidizer |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...