di-Ammonium oxalate monohydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Ammonium oxalate monohydrate, Ammonium oxalate-1-hydrate, Oxalic acid diammonium salt monohydrate, Oxalic acid ammonium salt monohydrate, Ammonium ethanedioate monohydrate, Ammonium Oxalate, Monohydrate

1.01192.0250 1.01192.1000
CA1.01192.0250CS 927 CAD
CA1.01192.0250 CA1.01192.1000
di-Ammonium oxalate monohydrate, EMSURE® for analysis, Supelco®
di-Ammonium oxalate monohydrate

1.01192.0250 will replace and is the same as AX1325-2. Part # Change only, Same Pack Size, Same Specifications.
1.01192.1000 replaces and is the same as AX1325-1. Different Pack Size, Same Specifications.

Formula: (NH₄)₂C₂O₄·H₂O
Boiling Pt: 70 °C (1013 hPa)
Melting Pt: 70 °C
Density: ∼1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149694
CAS Number: 6009-70-7
EINECS: 214-202-3
UN: 3288
ADR: 6.1,III
Merck Index: 13,00541

Order Now

Specification Test Results

Solubility 45 g/l (20 °C) (anhydrous substance)
Melting Point 70 °C (decomposition)
Molar Mass 142.11 g/mol
Bulk Density 480 kg/m3
Vapor Pressure (20 °C)
Density 1.50 g/cm3 (20 °C)
pH 6.3 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR