Diammonium hydrogen citrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.01154.0500 1.01154.2500
CA1.01154.0500EA 272 CAD
CA1.01154.0500 CA1.01154.2500
Diammonium hydrogen citrate, EMSURE® for analysis, Supelco®
di-Ammonium hydrogen citrate

EM1.01154.0500 replaces and is the same as AX1285-1; part# change only, Same Pack Size, Same Specifications.
EM1.01154.0500 also replaces and is the same as AX1285-11; different Pack Size, Same Specifications.
EM1.01154.2500 replaces and is the same as AX1285-3; part# change only, Same Pack Size, Same Specifications.
EM1.01154.9050 replaces and is the same as AX1285-20; part# change only, Same Pack Size, Same Specifications.

Formula: HOC(CO₂H)(CH₂CO₂NH₄)₂
Density: 1.483 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013068
CAS Number: 3012-65-5
EINECS: 221-146-3
Merck Index: 13,00513

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 226.19 g/mol
Bulk Density 400 - 600 kg/m3
Boiling Point (1013 hPa) (decomposition)
Vapor Pressure (20 °C)
Density 1.48 g/cm3 (20 °C)
pH 5.2 (50 g/l, H2O, 20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR