Ammonium chloride ≥99.8% (silver nitrate titration) [99.0-100.5% dry basis (silver nitrate titration)], EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Ammonium muriate, Salmiac

1.01145.0500 1.01145.5000 1.01145.1000 1.01145.9025 1.01145.9050
CA1.01145.0500CS 750 CAD
CA1.01145.0500 CA10754-008 CA1.01145.1000 CA1.01145.9025 CA1.01145.9050
Ammonium chloride ≥99.8% (silver nitrate titration) [99.0-100.5% dry basis (silver nitrate titration)], EMSURE® for analysis, Supelco®
Ammonium chloride
Formula: NH₄Cl
MW: 53.49 g/mol
Melting Pt: 335 (subl.)
Density: 1.53 g/cm3 (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 372 g/l (20 °C)
Melting Point 335 °C (sublimed)
Molar Mass 53.49 g/mol
Bulk Density 500 kg/m3
Vapor Pressure 1.3 hPa (30 °C)
Density 1.52 g/cm3 (20 °C)
pH 4.5 - 5.5 (50 g/l, H2O, 20 °C)
Ignition Point 400 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR