Ammonium chloride ≥99.995%, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Ammonium muriate, Salmiac

M-Clarity™ quality level = MQ100

1.01143.0050 1.01143.0500
CA1.01143.0050EA 269 CAD
CA1.01143.0050 CA1.01143.0500
Ammonium chloride ≥99.995%, Suprapur® for trace metal analysis, Supelco®
Ammonium chloride
Formula: NH₄Cl
MW: 53.49 g/mol
Melting Pt: 335 °C (subl., lit.)
Density: 1.53 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (metallic) ≥ 99.995 %
Al (Aluminium) ≤ 0.050 ppm
Ba (Barium) ≤ 5.0 ppm
Ca (Calcium) ≤ 0.50 ppm
Cd (Cadmium) ≤ 0.010 ppm
Co (Cobalt) ≤ 0.010 ppm
Cu (Copper) ≤ 0.010 ppm
Fe (Iron) ≤ 0.10 ppm
K (Potassium) ≤ 5.00 ppm
Li (Lithium) ≤ 0.10 ppm
Mg (Magnesium) ≤ 0.050 ppm
Mn (Manganese) ≤ 0.010 ppm
Na (Sodium) ≤ 1.0 ppm
Ni (Nickel) ≤ 0.010 ppm
Pb (Lead) ≤ 0.010 ppm
Sr (Strontium) ≤ 0.50 ppm
Tl (Thallium) ≤ 0.010 ppm
Zn (Zinc) ≤ 0.010 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR