Ammonium bromide ≥99.5% (silver nitrate titration), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.01125.1000 1.01125.9025
CA1.01125.1000EA 218 CAD
CA1.01125.1000 CA1.01125.9025
Ammonium bromide ≥99.5% (silver nitrate titration), EMSURE® for analysis, Supelco®
Ammonium bromide
Formula: H₄BrN
Melting Pt: >370 °C (closed capillary tube)
Density: 2.46 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00011419
CAS Number: 12124-97-9
EINECS: 235-183-8
Merck Index: 13,00505

Order Now

Specification Test Results

Solubility 742 g/l (20 °C)
Melting Point 542 °C (closed capillary tube)
Molar Mass 97.94 g/mol
Bulk Density 1100 kg/m3
Vapor Pressure 1.3 hPa (198 °C)
Density 2.43 g/cm3 (20 °C)
pH 4.8 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR