Nitric acid 65%, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.00456.1000 1.00456.2500 1.00456.2510
CA1.00456.1000CS 590 CAD
CA1.00456.1000 CA1.00456.2500 CA1.00456.2510
Nitric acid 65%, EMSURE® for analysis, Supelco®
Nitric acid
Formula: HNO₃
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 (5.1),II
Merck Index: 13,06608

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -32 °C
Boiling Point 121 °C (1013 hPa)
Vapor Pressure 9.4 hPa (20 °C)
Density 1.39 g/cm3 (20 °C)
pH (H2O, 20 °C) strongly acid
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR