Hydrofluoric acid 40%, Suprapur® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ100
1.00335.2500
1.00335.1000
1.00335.0250
1.00335.2500
1.00335.0250
1.00335.0500
1.00335.0500
1.00335.1000
CA1.00335.0500CS
2962.25
CAD
EM1.00335.1000
CA1.00335.0500
EM1.00335.0250
CA1.00335.2500
EM1.00335.2500
CA1.00335.0250
CA1.00335.1000
EM1.00335.0500
Hydrofluoric acid 40%, Suprapur® for trace metal analysis, Supelco®
Hydrofluoric acid
Formula:
HF MW: 20.01 g/mol |
MDL Number:
MFCD00011346 CAS Number: 7664-39-3 UN: 1790 ADR: 8,II |
Specification Test Results
Ag (Silver) | ≤ 0.5 ppb |
Al (Aluminium) | ≤ 2 ppb |
As (Arsenic) | ≤ 5 ppb |
Assay (acidimetric) | ≥ 40 % |
Au (Gold) | ≤ 0.2 ppb |
Ba (Barium) | ≤ 1 ppb |
Be (Beryllium) | ≤ 0.2 ppb |
Bi (Bismuth) | ≤ 0.2 ppb |
Ca (Calcium) | ≤ 5 ppb |
Cd (Cadmium) | ≤ 0.5 ppb |
Chloride (Cl) | ≤ 500 ppb |
Co (Cobalt) | ≤ 0.2 ppb |
Colour | ≤ 10 Hazen |
Cr (Chromium) | ≤ 1 ppb |
Cu (Copper) | ≤ 1 ppb |
Fe (Iron) | ≤ 5 ppb |
Ga (Gallium) | ≤ 0.2 ppb |
Ge (Germanium) | ≤ 1 ppb |
Hexafluorosilicate (SiF₆) | ≤ 20000 ppb |
Hg (Mercury) | ≤ 20 ppb |
In (Indium) | ≤ 0.2 ppb |
K (Potassium) | ≤ 10 ppb |
Li (Lithium) | ≤ 0.2 ppb |
Mg (Magnesium) | ≤ 2 ppb |
Mn (Manganese) | ≤ 2 ppb |
Mo (Molybdenum) | ≤ 0.2 ppb |
Na (Sodium) | ≤ 2 ppb |
Ni (Nickel) | ≤ 0.5 ppb |
Pb (Lead) | ≤ 0.5 ppb |
Pd (Palladium) | ≤ 0.2 ppb |
Phosphate (PO₄) | ≤ 100 ppb |
Pt (Platinum) | ≤ 0.2 ppb |
Residue on ignition (as sulphate) | ≤ 2000 ppb |
Sb (Antimony) | ≤ 0.2 ppb |
Sn (Tin) | ≤ 0.5 ppb |
Sr (Strontium) | ≤ 0.2 ppb |
Sulfite (SO₃) | ≤ 2000 ppb |
Sulphate (SO₄) | ≤ 500 ppb |
Ti (Titanium) | ≤ 5 ppb |
Tl (Thallium) | ≤ 0.2 ppb |
U (Uranium) | ≤ 0.1 ppb |
V (Vanadium) | ≤ 0.2 ppb |
Zn (Zinc) | ≤ 2 ppb |
Zr (Zirconium) | ≤ 0.2 ppb |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...