Hydrochloric acid 30%, Suprapur® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.00318.0250
1.00318.1000
1.00318.0500
CA1.00318.0250CS
617
CAD
CA1.00318.0250
CA1.00318.1000
EM1.00318.0500
Hydrochloric acid 30%, Suprapur® for trace metal analysis, Supelco®
Hydrochloric acid
Formula:
HCl Boiling Pt: 110 °C (1013 hPa) Melting Pt: –30 °C Density: 1.18 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Specification Test Results
Assay (acidimetric) | ≥ 30.0 % |
Colour | ≤ 10 Hazen |
Bromide (Br) | ≤ 10 ppm |
Free chlorine (Cl) | ≤ 300 ppb |
Phosphate (PO₄) | ≤ 10 ppb |
Sulphate (SO₄) | ≤ 300 ppb |
Sulfite (SO₃) | ≤ 500 ppb |
Ag (Silver) | ≤ 1.0 ppb |
Al (Aluminium) | ≤ 1.0 ppb |
As (Arsenic) | ≤ 1.0 ppb |
Au (Gold) | ≤ 1.0 ppb |
Ba (Barium) | ≤ 1.0 ppb |
Be (Beryllium) | ≤ 1.0 ppb |
Bi (Bismuth) | ≤ 1.0 ppb |
Ca (Calcium) | ≤ 5.0 ppb |
Cd (Cadmium) | ≤ 0.5 ppb |
Co (Cobalt) | ≤ 1.0 ppb |
Cr (Chromium) | ≤ 1.0 ppb |
Cu (Copper) | ≤ 1.0 ppb |
Fe (Iron) | ≤ 5.0 ppb |
Ga (Gallium) | ≤ 0.5 ppb |
Ge (Germanium) | ≤ 10.0 ppb |
Hg (Mercury) | ≤ 2.0 ppb |
In (Indium) | ≤ 0.5 ppb |
K (Potassium) | ≤ 10.0 ppb |
Li (Lithium) | ≤ 0.5 ppb |
Mg (Magnesium) | ≤ 2.0 ppb |
Mn (Manganese) | ≤ 0.5 ppb |
Mo (Molybdenum) | ≤ 0.5 ppb |
NH₄ (Ammonium) | ≤ 500 ppb |
Na (Sodium) | ≤ 10.0 ppb |
Ni (Nickel) | ≤ 1.0 ppb |
Pb (Lead) | ≤ 0.5 ppb |
Pt (Platinum) | ≤ 0.5 ppb |
Sb (Antimony) | ≤ 0.5 ppb |
Sn (Tin) | ≤ 1.0 ppb |
Sr (Strontium) | ≤ 0.5 ppb |
Ti (Titanium) | ≤ 1.0 ppb |
Tl (Thallium) | ≤ 0.5 ppb |
V (Vanadium) | ≤ 1.0 ppb |
Zn (Zinc) | ≤ 1.0 ppb |
Zr (Zirconium) | ≤ 0.5 ppb |
Residue on ignition (as sulphate) | ≤ 2 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...