Acetic acid glacial ≥100%, Suprapur® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.00066.0250
1.00066.1000
CA1.00066.0250CS
2781
CAD
CA1.00066.0250
CA1.00066.1000
Acetic acid glacial ≥100%, Suprapur® for trace metal analysis, Supelco®
Acetic acid
Formula:
H₃CCOOH Boiling Pt: 118 °C (1013 hPa) Melting Pt: 17 °C Density: 1.05 g/cm³ (20 °C) Flash Pt: 38.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 EINECS: 200-580-7 UN: 2789 ADR: 8 (3),II REACH: 01-2119475328-30 |
|
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 17 °C |
Molar Mass | 60.05 g/mol |
Boiling Point | 116 - 118 °C (1013 hPa) |
Vapor Pressure | 15.4 hPa (20 °C) |
Refractive Index | 1.37 (20 °C) |
Explosion Limit | 4 - 19.9 %(V) |
Density | 1.05 g/cm3 (20 °C) |
pH | 2.5 (50 g/l, H2O, 20 °C) |
Ignition Point | 485 °C |
Saturation concentration | 38 g/m3 (20 °C) Air |
Viscosity kinematic | 1.17 mm2/s (20 °C) |
Evaporation number | 24 |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...