Baffle for Temperature Probe, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
12193-03 12193-07 12193-09
89427-774EA 1527.24 CAD
89427-774 89427-776 89427-778
Baffle for Temperature Probe, PTFE, Ace Glass Incorporated
Stirrers Stirring Tools
A PTFE baffle with an integral stainless steel tube
Can be used for either liquid addition into a reactor or primarily a temperature probe. The baffle will accomodate a 1/4" OD thermocouple. The #7 Ace-Thred bottom bushing will make a tight compression seal with either the tubing or thermocouple. Thermocouple not included.