Vacuum Trap, Two-Piece, Bent Vertical Tube, Ace Glass Incorporated
Supplier: Ace Glass
8760-12 8760-18 8760-24 8760-64 8760-74 8760-06 8760-08 8760-10 8760-16 8760-22 8760-62 8760-72
89085-076EA 388.49 CAD
89085-076 89085-078 89085-080 89085-082 89085-084 89235-716 89235-718 10099-506 10099-508 10099-510 10099-512 10099-514
Vacuum Trap, Two-Piece, Bent Vertical Tube, Ace Glass Incorporated
Vacuum Traps
Two piece vacuum trap design facilitates easy cleaning
Stopper features hose connection ends and a bent vertical tube. The trap bodies only are available separately.