Vacuum Trap, Round Bottom Flask with Adjustable Inner Tube, Ace Glass Incorporated
Supplier: Ace Glass
8744-06 8744-08 8744-09 8744-10 8744-11 8744-13 8744-14 8744-20 8744-23 8744-25
89063-796EA 301.31 CAD
89063-796 89063-798 89063-800 89063-802 89063-804 89063-806 89063-808 89063-810 89063-812 89063-814
Vacuum Trap, Round Bottom Flask with Adjustable Inner Tube, Ace Glass Incorporated
Vacuum Traps
Round bottom with adjustable inner tube and nylon bushing with FETFE o-ring
Optional threaded adapters will allow safe and easy tubing connect-disconnect. Simply attach tubing to serrated end of adapter, then connect adapter via a bushing to inlet and outlet of trap. Serrated end of adapter has six rings; smallest 14 mm O.D., largest 16 mm O.D. Adapter not supplied with complete unit.