Vacuum Manifold with Firestone Rapid Purge Valve and Easy Action Stopcocks, Ace Glass Incorporated
Supplier: Ace Glass
8765-13 8765-15 8765-27 8765-29 8765-34 8765-36
89064-012EA 2552.56 CAD
89064-012 89064-014 89064-016 89064-018 89064-020 89064-022
Vacuum Manifold with Firestone Rapid Purge Valve and Easy Action Stopcocks, Ace Glass Incorporated
Manifolds
Vacuum/gas manifold with a Firestone rapid purge valve at one end and 0–5 mm high vacuum, Easy-Action PTFE stopcock ports with 3/8" O.D
Swagelok tubing connections. Each connection has an o-ring groove (o-rings supplied) for use with #11 Ace-Thred. Valve eliminates the need for a second tube. Simply connect one 10 mm O.D. arm to vacuum, other to gas source, and by half turn of the stopcock plug you can alternate between vacuum and gas. Shut-off float on gas side of valve prevents pressure buildup when system is purged. Plugs are PTFE with UHDPE or PTFE handles with FETFE o-rings.
Ordering information: For replacement plugs, see 89062-486(UHDPE) or 89062-578(PTFE).