Addition Funnels, Graduated with Glass Plug, Ace Glass Incorporated
Supplier: Ace Glass
7267-16 7267-12 7267-20 7267-18 7267-08
89058-546EA 762.01 CAD
89058-546 89058-652 89058-676 89058-722 89058-768
Addition Funnels, Graduated with Glass Plug, Ace Glass Incorporated
Funnels Dropping Funnels
Cylindrical addition funnel with [ST] joint top and bottom, drip stem, double-scale graduated and calibrated up from the stopcock
The drip stem does not extend beyond the joint, thereby eliminating a possible source of breakage. Stopcock is available with 1:5 PTFE plug, PTFE metering plug, or glass plug
Ordering information: For stoppers (not supplied), order 89062-824 (24/40) or 89062-850 (14/20). For replacement 1:5 PTFE stopcock plugs, order 89062-770 (2-bore) or 89062-774 (4-bore). For replacement metering valves, order 89062-806 (2 mm) or 89062-808 (4 mm). For replacement glassMore Product Information