Glass Check Valve with Ground Seat for Liquid Seal, Ace Glass Incorporated
Supplier: Ace Glass
8770-05 8770-10
89064-038EA 261.79 CAD
89064-038 89064-040
Glass Check Valve with Ground Seat for Liquid Seal, Ace Glass Incorporated
Valves Vacuum Valves
An all-glass check valve with ground seat for liquid seal
Available in style A — for vertical use, and style B — for right angle connection. Connections are 8 mm O.D.