Separatory Funnel, European Style, Externally Threaded, Body Only, Ace Glass Incorporated
Supplier: Ace Glass
7231-44 7231-34 7231-36 7231-38 7231-40 7231-42
89058-576EA 1049.05 CAD
89058-576 89058-578 89058-580 89058-582 89058-584 89058-586
Separatory Funnel, European Style, Externally Threaded, Body Only, Ace Glass Incorporated
Funnels Separating Funnels
European style funnel, body only
Outer ground joint at top is a Rodaviss (externally threaded) joint. Stopper can be secured in top joint via cap and o-ring for a positive, leak-tight seal. Leak-tight, locking stopper available separately. Fabricated from borosilicate glass.