Separatory Funnel, European Style, with PTFE Stopcock and Polyethylene Stopper, Ace Glass Incorporated
Supplier: Ace Glass
7231-06 7231-09 7231-12 7231-15 7231-18 7231-21
89058-566EA 345.65 CAD
89058-566 89058-568 89058-570 89058-572 89058-574 89058-588
Separatory Funnel, European Style, with PTFE Stopcock and Polyethylene Stopper, Ace Glass Incorporated
Funnels Separating Funnels
European style funnel
Supplied with a standard taper 24/25 polyethylene stopper in all capacities except the 2000 mL size which is supplied with a large standard taper 45/50 polyethylene stopper. Fabricated from borosilicate glass.