Kord-Valmark Disposable Polystyrene Petri Dishes, Slippable, Akro-Mils
Supplier: BIO PLAST MANUFACTURING
42529020000100 42529040000750 42529050000400 42529090000340 42529100000500 4252910000500P 42529110000500 42529140000500 42529160000500 42529170000500
CA11019-580CS 143.15 CAD
CA11019-580 11019-554 11019-558 11019-560 11019-564 11019-566 11019-568 11019-570 11019-576 11019-580 11019-582 11019-588
Kord-Valmark Disposable Polystyrene Petri Dishes, Slippable, Akro-Mils
Culture Dishes Petri Dishes
Kord-Valmark by Akro-Mils disposable polystyrene petri dishes are optically clear and radiation sterilized
Slippable polystyrene petri dishes are recommended for standard microbiology and general laboratory use, including the use of automated filling and streaking systems. Slippable lids provide a semi-stacking feature for hand use, while ensuring ease of use in automated equipment. Automation lids have no rim, ensuring ease of use in automated equipment. Select products are available with Side Arrows and/or ISO Mark Targets. Slippable petri dishes are available in 60 x 15 mm, 100 x 15 mmMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable