Separatory Funnel, Globe Style with Glass Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
7224-22 7224-28 7224-33
89058-514EA 1195.65 CAD
89058-514 89058-516 89058-518
Separatory Funnel, Globe Style with Glass Stopcock, Ace Glass Incorporated
Funnels Separating Funnels
This is a separatory, globe-type funnel with a neck and a section between the globe and the stopcock shaped to provide a secure grip for shaking contents or transporting
Also makes clamping easier. Fabricated from borosilicate glass.