Kord-Valmark Disposable Polystyrene Petri Dishes, Stackable, Akro-Mils
Supplier: BIO PLAST MANUFACTURING
42529000000500 42529010000500 42529030000500 42529060000340 42529120000500 42529130000500
11019-550CS 387.18 CAD
11019-550 11019-552 11019-556 11019-562 11019-572 11019-574
Kord-Valmark Disposable Polystyrene Petri Dishes, Stackable, Akro-Mils
Culture Dishes Petri Dishes
Kord-Valmark by Akro-Mils disposable polystyrene petri dishes are optically clear and radiation sterilized
Stackable polystyrene petri dishes are recommended for standard microbiology or general laboratory use. The stackable lid design ensures stack stability and clear top and bottom viewing surfaces. Stackable dishes are available in 60 x 15 mm, 100 x 15 mm (Mono, Bi, Tri and Quad plate) and 100 x 25 mm sizes.
Kord-Valmark by Akro-Mils Petri Dishes are sterile if sleeves are unopened. Ventilation (3, 4 OR 8) ribs reduce condensation and allow air circulation. Consistent flatness allows for even media distribution. Flared lid skirt and squared corners allow for easy one-hand operation, especially with gloves. Product is Traceable by lot and SKU number. Certificates of Compliance are available upon request.
Ordering information: Accessories not included.