LeStreak® Microstreakers, Decon Labs

Supplier: Decon Labs
MP1855 MP1875 MP1845 MP1825 MP1805
CA11027-268PK 1283 CAD
CA11027-268 CA11027-270 CA11027-272 CA11027-274 CA11027-276
LeStreak® Microstreakers, Decon Labs
Spreaders Inoculating Loops and Needles
LeStreak® Microstreakers are used to inoculate or transfer microbiological samples

  • Small LeStreak® loop: Int.Ø 2.26 mm

  • Medium LeStreak® loop: Int.Ø 2.91 mm
  • Large LeStreak® loop: Int.Ø 3.57 mm


Decon’s LeStreak® microstreakers are loops made of high-quality, twisted nichrome wire that is permanently attached to lightweight, insulated handles which are perfectly balanced for easy manipulation. LeStreak® microstreakers slide smoothly across the agar surface with out tearing or gouging the medium. The nichrome wire cools rapidly after flaming.



The 1 and 10 µl (0.001 and 0.01 ml)) calibrated inoculating LeStreakers are individually calibrated in accordance with ASM/APHA via Evans blue dye test which is a CAP approved method. Calibration certificates are sent with calibrated loops or are available from Decon labs.



Certifications: UL listed.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR