OdorGone® Autoclave Deodorant Capsules, DLI, Decon Labs
Supplier: Decon Labs
LEM100 LEM25 APP25 LEM25 APP100 APP100 LEM100
CA11027-244CS 2648.75 CAD
89426-258 CA11027-244 CA11027-240 89426-264 89426-262 89426-260 CA11027-239
OdorGone® Autoclave Deodorant Capsules, DLI, Decon Labs
Deodorants and Air Fresheners
OdorGone® Autoclave Deodorant Capsules are activated by the heat and moisture from the autoclave to release the contents of the capsule and overcome unpleasant smells generated from heating microbiological waste
One capsule per load absorbs offensive odors leaving the laboratory smelling fresh and clean. Available in apple or lemon frangrance.
Certifications: UL listed.