Single Neck Vacuum Flasks with Side Arm, Ace Glass Incorporated
Supplier: Ace Glass
7003-11 7003-13 7003-15 7003-25 7003-27 7003-29 7003-211 7003-213 7003-215 7003-217 7003-219
89428-034EA 354.91 CAD
89428-034 89428-036 89428-038 89428-040 89428-042 89428-044 89428-046 89428-048 89428-050 89428-052 89428-054
Single Neck Vacuum Flasks with Side Arm, Ace Glass Incorporated
Flasks Round Bottom Flasks
Round bottom flask with a single neck with standard taper joint and an 8 mm side outlet with either a 2 mm glass stopcock or a 0-3 inlet valve
Ideal for Schlenk lines and either NO-AIR or vacuum applications. Fabricated from borosilicate glass. Includes one 89064-960 septa for side arm.