Filtration Apparatus, Ace Glass Incorporated
Supplier: Ace Glass
3708-02 3708-10 3708-20 3709-05 3709-18 3708-14 3708-16 3709-16
89427-882EA 3264.41 CAD
89427-882 89427-884 89427-886 89427-888 89427-890 89428-174 89428-176 89428-178
Filtration Apparatus, Ace Glass Incorporated
Filters Membrane Filter Units
This borosilicate glass filtration apparatus accomodates 75 mm membrane filters.
The new design utilizes a PTFE Ace-Thred adapter to join the glass funnel, filter flask and fritted disc together. This design eliminates the need for the clamping device, stopper and base. The unit is easy to assemble, disassemble and clean. The fritted disc can be easily replaced and other porosity discs used. The apparatus is easier to use and more economical than old designs.