Reflux Distillation Splitter, Ace Glass Incorporated
Supplier: Ace Glass
6088-08 6088-10 6088-12 6088-14
89427-902EA 1095.36 CAD
89427-902 89427-904 89427-906 89427-908
Reflux Distillation Splitter, Ace Glass Incorporated
Distillation Components Distillation Sets
Reflux distillation splitter adapter for all sizes of reactors
Standard taper inner joint bottom and reinforced outer joint top. A slight angle to the bottom joint allows for addition to reactor heads with angled side joints. Alignment allows for straight flow path up- wards to the condenser. Three standard joint sizes are available. Side valve is 0-10mm with a precision ground bottom take off that matches 1/2" compression fittings. Valve replacement FETFE o-rings are sizes -011 and -111. Fabricated from borosilicate glass.