Graduated Vacuum Storage Tube with Inlet Valve and Inner Joint Sidearm, Ace Glass Incorporated
Supplier: Ace Glass
7756-217 7756-223 7756-211
89084-866EA 408.77 CAD
89084-866 89084-872 89084-882
Graduated Vacuum Storage Tube with Inlet Valve and Inner Joint Sidearm, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
This tube is used for carrying out most reactions in solution with filter tube
Also may be used with a finger condenser in center joint for reflux reactions. Available with either a stopcock or valve side port and a standard taper 14/35 or standard taper 24/40 side joint, and either a standard taper 14/20 or standard taper 24/25 outer center joint opening. Graduated. Fabricated from borosilicate glass. Normal usage requires 89077-938 stirring magnet.
Order Now
SPECIFICATIONS
Environmentally Preferable