Addition Funnel, Jacketed, Ace Glass Incorporated
Supplier: Ace Glass
7278-15 7278-17 7278-07 7278-25 7278-23 7278-11 7278-19
89428-064EA 1539.68 CAD
89428-064 89428-062 89428-058 89428-068 89428-066 89428-060 89428-070
Addition Funnel, Jacketed, Ace Glass Incorporated
Funnels Dropping Funnels
Graduated addition funnels with outside jacket for cooling and/or heating
Jacket extends from just below shoulder of vessel down to the bottom tube just below the stopcock. Stopcock is either 2 or 4 mm bore PTFE plug. Bottom drip tube extends to the edge of the lower, inner standard taper, joint. Top outer reinforced joint size matches the bottom joint size. Side hose barbs are size D for 3/8-in ID tubing. Fabricated from borosilicate glass.