Distillation Packing, Ace Glass Incorporated
Supplier: Ace Glass
6624-04 6624-09 6624-23 6624-28
89428-180EA 976.64 CAD
89428-180 89428-182 89428-184 89428-186
Distillation Packing, Ace Glass Incorporated
Distillation Column Packing
An efficient protruded metal dump-packing for distillation columns of 1" to 12" diameter
  • Self-wetting reduces equilibrium time
  • The dump feature eliminates interlocking, and makes filling and emptying as easy as pouring water
  • Suitable for both atmospheric and reduced pressures down to 10mm Hg condenser pressure
  • Packed heights up to 20 feet are possible without distorting packing
  • Metal packing offered in 316 Stainless Steel and Monel 400
Minimum column diameter for 0.16" packing is 0.75".