24/40 Powder Dispensing Funnel, Auger Style, Ace Glass Incorporated
Supplier: Ace Glass
7239-40 7239-30
89058-608EA 1165 CAD
89058-608 89058-640
24/40 Powder Dispensing Funnel, Auger Style, Ace Glass Incorporated
Funnels Powder Funnels
Side operated screw feeds funnel for the addition of powders and solids (up to 25 mesh) to reactions without problems of seizing or binding
PTFE-ceramic shaft and contained within a precision bore housing. This offers a unique flexibility in that the thread can move aside temporarily; as one spacing increases, the adjacent spacing decreases, thereby maintaining an average feed rate. Completely inert materials allow flushing with solvents without fear of contamination.
Standard taper 14/20 outer joint atop bottom inner joint has been eliminated. Bottom standard taper joint and top standard taper outer joints are 24/40. Features a flexible PTFE screw thread wrapped on a precision. Fabricated from borosilicate glass.