Addition Funnels with 1:5 PTFE Plug, Ace Glass Incorporated
Supplier: Ace Glass
9498-05 9498-11 9498-03
89074-994EA 443.8 CAD
89074-994 89074-996 10099-114
Addition Funnels with 1:5 PTFE Plug, Ace Glass Incorporated
Funnels Dropping Funnels
Ungraduated with a standard taper joint top and bottom
Fabricated from borosilicate glass. For stoppers, see Round-Head Glass Stoppers, Ace Glass Incorporated.