24/40 Moisture Test Receiver, Modified Bidwell and Sterling with Overflow, Ace Glass Incorporated
Supplier: Ace Glass
7735-02 7735-04
89060-322EA 502.55 CAD
89060-322 89060-330
24/40 Moisture Test Receiver, Modified Bidwell and Sterling with Overflow, Ace Glass Incorporated
Distillation Components Distillation Receivers
Modified Bidwell & Sterling type with overflow to allow excess solvent to return to boiling flask
Both capacities are calibrated in 0.1 mL subdivisions. Joints are a standard taper 24/40.