Hi-Recovery Vials, Ace Glass Incorporated
Supplier: Ace Glass
5703-01 5703-02 5703-04 5703-05 5703-10 5703-20
89427-714CS 705.85 CAD
89427-714 89427-716 89427-718 89427-720 89427-722 89427-724
Hi-Recovery Vials, Ace Glass Incorporated
Vials
Clear type 1 borosilicate glass vials with recessed bottom for both concentration of material during reactions, extractions and chromatography applications.
Vials are either straight sided with a conical bottom or a tapered internal bottom.