Headspace Vial Threaded Magnetic Caps, 18 mm, Ace Glass Incorporated
Supplier: Ace Glass
5710-05 5710-07 5710-09 5710-11
89427-810PK 313.37 CAD
89427-810 89427-812 89427-814 89427-816
Headspace Vial Threaded Magnetic Caps, 18 mm, Ace Glass Incorporated
Closures Chromatography Vial Closures
These are magnetic, aluminum, 18 mm thread caps for headspace vials
They work with all autosamplers. The caps have a bonded PTFE/Silicone liner.