UniRack™ and UniRack™Jr. Centrifuge Tube Racks, Simport Scientific
Supplier: Simport Scientific
S500-80B S500-80G S500-80O S500-80P S500-80R S500-80Y S500-25B S500-25R S500-25Y
CA60873-002CS 86.66 CAD
CA60873-002 CA60873-004 CA60873-006 CA60873-008 CA60873-010 CA60873-012 CA89188-002 CA89188-004 CA89188-006
UniRack™ and UniRack™Jr. Centrifuge Tube Racks, Simport Scientific
Racks Tube Racks
The UniRack™ can hold up to 25 polystyrene or polyprolylene 10 and 12 mm tubes
This rack will accommodate all types of screw cap microtubes from 0.5 to 2 mL as well as 1 to 5 mL cryogenic vials. Flip the UniRack™ over and you can store up to 16 PCR or microcentrifuge tubes from 0.2 to 0.5 mL. Supplied without handles or anchor pins.