Aluminum Seals, 20 mm, Lined, Ace Glass Incorporated
Supplier: Ace Glass
5708-05 5708-06 5708-10 5708-11 5708-12 5708-18 5708-19 5708-20 5708-30 5708-31 5708-32 5708-33 5708-34 5708-35 5708-36 5708-37 5708-38
89097-124CS 3089.68 CAD
89097-124 89097-126 89097-128 89097-130 89097-132 89097-134 89097-136 89097-138 89427-792 89427-794 89427-796 89427-798 89427-800 89427-802 89427-804 89427-806 89427-808
Aluminum Seals, 20 mm, Lined, Ace Glass Incorporated
Closures Chromatography Vial Closures
These seals are 20mm lined, natural aluminum crimp style seals for headspace, serum and other styles of vials with 20mm crimp finish tops
Available in natural aluminum with various PTFE/rubber linings. Colored aluminum available via special order. A magnetic crimp seal version is available for use with robotics and a pressure release version designed to release at 3 Bar (43 psi) pressure.