Pressure Equalizing Funnel, Graduated, with 1:5 PTFE Plug, Ace Glass Incorporated
Supplier: Ace Glass
7286-08 7286-12 7286-10 7286-14
89058-542EA 907.38 CAD
89058-542 89058-666 89058-672 89058-708
Pressure Equalizing Funnel, Graduated, with 1:5 PTFE Plug, Ace Glass Incorporated
Funnels Separating Funnels
Cylindrical with standard taper joint at top and bottom and a drip stem
Stopcock on equalizing arm permits filling funnel without having to open the system to atmosphere. For stoppers, order 89062-824. Stopper not included.