Plates, Laser Coded, Silica Gel 60, MilliporeSigma
Supplier: MilliporeSigma
1.05566.0001 1.05613.0001 1.05564.0001 1.05566.0001 1.05613.0001 1.05564.0001
CA1.05566.0001EA 1664.56 CAD
CA1.05566.0001 CA1.05613.0001 EM1.05564.0001 EM1.05566.0001 EM1.05613.0001 EM1.05702.0001 CA1.05564.0001 CA1.05702.0001
Plates, Laser Coded, Silica Gel 60, MilliporeSigma
TLC Plates
These plates feature individual laser coding for GLP applications
Product # 1.05566.0001 replaces and is identical to Product # 105566-7.
Product # 1.05613.0001 replaces and is identical to Product # 5613-6.
Designed for GLP applications, the tops of each of these plates bear an item, batch, and individual plate number. As a result, lab professionals can easily record and archive every plate they use. Based on proven silica found in TLC and HPTLC plates, these GLP plates deliver equally unsurpassed separation performance.