Tamper-Evident Micrewtube® Microcentrifuge Tube with Screw Cap, Simport Scientific
Supplier: Simport Scientific
T335-2TP T335-2STP T335-2SPRTP T335-4TP T335-4STP T335-4SPRTP T335-5TP T335-5STP T335-5SPRTP T335-6TP T335-6STP T335-7TP T335-7STP T335-7SPRTP T335-2TP T335-2STP T335-2SPRTP T335-4TP T335-4STP T335-4SPRTP T335-5TP T335-5STP T335-5SPRTP T335-6TP T335-6STP T335-7TP T335-7STP T335-7SPRTP
89499-730CS 1263 CAD
89499-730 89499-732 89499-734 89499-736 89499-738 89499-740 89499-742 89499-744 89499-746 89499-748 89499-750 89499-752 89499-754 89499-756 CA11033-378 CA11033-380 CA11033-382 CA11033-384 CA11033-386 CA11033-388 CA11033-390 CA11033-392 CA11033-394 CA11033-396 CA11033-398 CA11033-402 CA11033-404 CA11033-406
Tamper-Evident Micrewtube® Microcentrifuge Tube with Screw Cap, Simport Scientific
Tubes Microcentrifuge Tubes
A Simport MICREWTUBE® ideal for applications requiring a tamper-evident seal, as well as for transport, freezer storage, boiling application and centrifugation.
The screw cap features an innovative tamper-evident locking ring, leakproof O-ring seal, and a flat top for writing and packaging applications. The cap is molded with a deep internal lip that fits snugly against the interior wall of the tube thus preventing the contents from coming in contact with the seal or threads. Tubes and caps are made of polypropylene. Will withstand temperatures from –196 to +121 °C and are autoclavable. Non skirted tubes withstand high-speed centrifugation up to 20000×g, skirted tubes up to 17000×g.
These tubes fit most standard microcentrifuge rotors. Three sizes of conical bottom or self-standing tubes (0.5, 1.5 and 2 ml) are available in sterile and non-sterile format. All sterile tubes are gamma irradiated and packaged in tamperproof, resealable bags to protect remaining tubes from contamination.
Certifications: Certified RNase-, DNase-, DNA-, and pyrogen-free.
Caution: Do not use for storage in the liquid phase of liquid nitrogen.