Non-Graduated Transfer Pipettes, Globe Scientific
Supplier: Globe Scientific
138020B 138030 138040 138050 138060 138090 138060-S20 138090-S01 138040-S01 138040-S20 138080-S01 138080-S20 138020-S01 138020-S20 138030-S01 138030-S20 138050-S01 138050-S20 138060-S01
89497-692CS 1061.23 CAD
89497-692 89497-694 89497-696 89497-698 89497-700 89497-704 76285-364 76285-366 76285-372 76285-374 76285-376 76285-378 76285-390 76285-392 76285-394 76285-396 76285-398 76285-400 76285-402
Non-Graduated Transfer Pipettes, Globe Scientific
Pipettes Pasteur/Transfer Pipettes
Produced from a custom grade low-density polyethylene (LDPE), these unbreakable one-piece pipettes are used to draw and dispense liquids in controlled, reproducible drop sizes.
  • Non-toxic and chemically inert to biological fluids and most acids
  • Easy to grip with gloves
  • Temperature Range: –50 to +90 °C
  • Can be gas (EtO) sterilized
Offered in a variety of styles, these popular non-graduated general purpose transfer pipettes are packaged in a unique dispenser box. The convenient box is small, compact, and ideal for benchtop use. The hinged front panel opens for quick access and can be closed when not in use to keep the contents clean. Each dispenser box contains 500 pipettes.