Pressure Equalizing Funnel, Cylindrical, Ace Glass Incorporated
Supplier: Ace Glass
7285-35 7285-37 7285-39
89058-696EA 827.86 CAD
89058-696 89058-698 89058-700
Pressure Equalizing Funnel, Cylindrical, Ace Glass Incorporated
Funnels Separating Funnels
Cylindrical, with standard taper 24/40 joint at top and bottom
Supplied with 1:5 PTFE stopcock plugs which require no lubrication. Stopcock on equalizing arm permits filling funnel without having to open the system to atmosphere. Drip tip does not extend beyond joint, thereby eliminating a possible source of breakage. For stoppers, order 89062-824. Stopper not included.