Optimize Back Pressure Regulators
Supplier: Optimize
10-06-00120 10-06-00128 10-06-00130 10-06-00132 10-06-00129
21510-684EA 867.64 CAD
21510-684 77100-074 77100-078 21508-953 89522-314
Optimize Back Pressure Regulators
Gas Regulators
Optimize flow-through back pressure regulators utilize an active mechanism to maintain steady back pressure across a range of mobile phase viscosities and flow rates.
  • Flow-through design
  • Utilizes an active mechanism to maintain steady back pressure across a range of conditions
  • Prevent solvent decompression and bubble formation in the flow cell
  • Reduces baseline noise and drift
  • 100 µl internal volume
Placing one downstream from your detector will prevent solvent decompression and bubble formation in the flow cell, reducing baseline noise and drift. Each pre-set Optimize stainless steel Back Pressure Regulator offers a 100 µl internal volume and includes fittings for immediate connection out of the box.
Delivery information: Includes OPTI-LOK® LP Fittings.