OPTI-LOK™ High Pressure Fittings
Supplier: Optimize
10-20-00272 10-20-00274 10-20-00276 10-20-00255 10-20-00256 10-20-00266 10-20-00251 10-20-00253 10-20-00258 10-20-00259 10-20-00263 10-20-00264 10-20-00265 10-20-00268 10-20-00269 10-20-00270 10-20-00271 10-20-00273 10-20-00275 10-20-00518 10-24-00367 10-24-00369 10-24-00370 10-24-00371 10-24-00373 10-24-00375 10-24-00376 10-20-00261
21510-159PK 655.79 CAD
21510-159 21510-163 21510-167 21510-680 21510-682 21510-696 77100-154 77100-156 77100-158 77100-160 77100-166 77100-168 77100-170 77100-172 77100-174 77100-176 77100-178 77100-180 77100-182 77100-188 77100-260 77100-264 77100-266 77100-268 77100-270 77100-272 77100-274 89522-310
OPTI-LOK™ High Pressure Fittings
Fittings and Connectors
OPTI-LOK Fittings are ideal for high-pressure connections with ¹/₁₆" outer Ø tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6000 psi.
  • Precision machined for optimal quality and thread consistency
  • Zero-dead-volume connections every time
  • Trouble-free operation at high pressures
We realize that your time is valuable and should not be wasted worrying about fittings. For this reason, all Optimize fittings are precision machined in the USA for optimal quality and thread consistency. The machining process is the best way to ensure tight manufacturing tolerances and minimal product variance. What does all this mean? It means that every fitting you purchase from Optimize will perform just as well as the last, so you'll have no worries about faulty fittings. This reflects our commitment to providing our customers with the highest quality, best designed product available.
Opti-Lok LP Fittings are an excellent choice for making low-pressure connections into ¹/₄-28 ports. Connections to flat-bottomed ports can be made using Opti-Lok flat-bottomed ferrules for ¹/₁₆ or ¹/₈" tubing, without the need for pre-flanged tubing ends. Opti-Lok LP fittings are precision machined to deliver leak-free. Also available are ¹/₄-28 unions in various through-hole sizes for joining flatbottom fittings together.
Opti-Lok Adapters offer maximum flexibility for making connections between your ¹/₄-28, M6 (Metric), Luer and 10-32 fittings and ports. They are available in a useful variety of thread-type combinations, all precision machined from PEEK.
Ordering information: Available individually and in packs of ten.