Mat Covers for BioBlock™ Deep Well Plates, Simport Scientific
Supplier: Simport Scientific
T110-25 T110-26 T110-25 T110-26
CA48648-155CS 286.46 CAD
CA48648-155 CA48648-156 89499-396 89499-398
Mat Covers for BioBlock™ Deep Well Plates, Simport Scientific
Seals Microplate Seals
Designed to fit Bioblock™ Deep Well Plates, these mat covers are made of a specially formulated plastic ensuring great flexibility.
  • Allow for maximum sample volume in each well
  • Resistant to DMSO and biological buffers
Covers 89499-396 and 89499-398 are made of polyolefin and elastomer. These covers are recommended for use within a temperature range of –80 to 80°C.