Funnel-Reservoir, Ace Glass Incorporated
Supplier: Ace Glass
7342-18 7342-14 7342-16 7342-12
89058-658EA 2312.52 CAD
89058-658 89058-714 89058-724 89058-770
Funnel-Reservoir, Ace Glass Incorporated
Funnels Filtration Funnels Filter Funnels
Cylindrical, graduated reservoir permits determination of volume added at any time during pumping cycle
By stopcock manipulation, material may be pumped from graduated (10 mL in 0.1 mL) side tube only. This permits obtaining accurate pumping rate over short periods of time. Top joint is standard taper 24/40 outer. Outside diameter of bottom outlet is 9.5 mm (3/8-inch) tubing which fits Swagelok.