Addition Funnels with Stopper, Ace Glass Incorporated
Supplier: Ace Glass
9486-03 9486-05
89074-934EA 633.66 CAD
89074-934 89074-936
Addition Funnels with Stopper, Ace Glass Incorporated
Funnels Dropping Funnels
With an equalizing arm and a special stopper so that material may be isolated from the atmosphere
Turning the stopper completely closes the vessel to the equalizing arm so that the funnel may be disconnected without exposing contents to moist air, etc. Glass stopcock is 2 mm bore.
Ordering information: For replacement stoppers, order 89074-938.