Flanged Plug Caps, 12 mm, Globe Scientific
Supplier: Globe Scientific
118127K 118127L 118127N 118127W
89497-868PK 53.62 CAD
89497-868 89497-870 89497-872 89497-874
Flanged Plug Caps, 12 mm, Globe Scientific
Closures Test/Sample Tube Closures
This line of polyethylene flanged plug caps provides exceptional leak-resistance in test tubes, centrifuge tubes, and round cuvettes.
  • Low density polyethylene (LDPE)
  • Assorted colors available
  • Fits both glass and plastic tubes
Flanged plug caps feature two flexible flanges that press firmly against the inside wall of the tubes, thereby providing better leak-resistance than standard plug caps. Available in a variety of colors to facilitate identification.
Order Now
SPECIFICATIONS
Environmentally Preferable