Dual Position Closure Caps, Globe Scientific
Supplier: Globe Scientific
118118 118120
89497-864PK 186.62 CAD
89497-864 89497-866
Dual Position Closure Caps, Globe Scientific
Closures Test/Sample Tube Closures
This dual position cap is produced from low density polyethylene (LDPE). It is ribbed for easy handling.
The cap is specially designed to provide aerobic or anaerobic storage of culture samples. When snapped into the tube, the cap provides a leak-resistant seal.
Note: Leave the cap in the loose position for aerobic work and in the sealed position for anaerobic cultures.