Flanged Plug Caps, 16 mm, Globe Scientific
Supplier: Globe Scientific
116152B 116152C 116152G 116152K 116152L 116152N 116152R 116152W 116152Y
89497-844PK 76.31 CAD
89497-844 89497-846 89497-848 89497-850 89497-852 89497-854 89497-856 89497-858 89497-860
Flanged Plug Caps, 16 mm, Globe Scientific
Closures Test/Sample Tube Closures
This line of polyethylene flanged plug caps provides exceptional leak-resistance in test tubes, centrifuge tubes, and round cuvettes.
  • Low density polyethylene (LDPE)
  • Assorted colors available
  • Fits both glass and plastic tubes
Flanged plug caps feature two flexible flanges that press firmly against the inside wall of the tubes, thereby providing better leak-resistance than standard plug caps. Available in a variety of colors to facilitate identification.